Search
Search
| Gene Symbol: ADAM8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018041616.1 | XP_017897105.1 | 4 - 5 | 432 | 58 | |
| XM_018041617.1 | XP_017897106.1 | 4 - 5 | 432 | 58 | ||
| XM_018041618.1 | XP_017897107.1 | 4 - 5 | 432 | 58 | ||
Set Membership: |
|