Search
Search

C11H2orf49| Gene Symbol: C11H2orf49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013967476.2 | XP_013822930.1 | 2 - 3 | 88 | ||
| GenBank mRNA | JT679776.1 | - | 3 - 4 | 538 | 88 | |
| JT740037.1 | - | 3 - 4 | 400 | 88 | ||
Set Membership: |
|