Search
Search

UBIAD1| Gene Symbol: UBIAD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005690683.3 | XP_005690740.1 | 1 - 2 | 866 | 83 | |
| GenBank mRNA | JO322504.2 | - | 1 - 2 | 435 | 83 | |
| JO436271.2 | - | 1 - 2 | 753 | 83 | ||
Set Membership: |
|