Search
Search

CCDC9B| Gene Symbol: CCDC9B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013966975.2 | XP_013822429.2 | 5 - 6 | 700 | 132 | |
| XM_018054452.1 | XP_017909941.1 | 4 - 5 | 1252 | 132 | ||
| XM_018054453.1 | XP_017909942.1 | 5 - 6 | 894 | 132 | ||
| XM_018054454.1 | XP_017909943.1 | 4 - 5 | 1514 | 132 | ||
| XM_018054455.1 | XP_017909944.1 | 3 - 4 | 508 | 132 | ||
Set Membership: |
|