Search
Search

LRRC8B| Gene Symbol: LRRC8B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018045726.1 | XP_017901215.1 | 4 - 5 | 2396 | 63 | |
| XM_018045727.1 | XP_017901216.1 | 4 - 5 | 2583 | 63 | ||
| GenBank mRNA | JO427051.1 | - | 4 - 5 | 2543 | 63 | |
Set Membership: |
|