Search
Search
| Gene Symbol: RBBP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005690394.3 | XP_005690451.1 | 3 - 4 | 371 | 63 | |
| XM_005690395.3 | XP_005690452.1 | 3 - 4 | 371 | 63 | ||
| XM_018060041.1 | XP_017915530.1 | 3 - 4 | 371 | 63 | ||
Set Membership: |
|