Search
Search
| Gene Symbol: KLF15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005696172.3 | XP_005696229.1 | 11 - 12 | 1354 | 91 | |
| XM_018067114.1 | XP_017922603.1 | 11 - 12 | 1191 | 91 | ||
| XM_018067115.1 | XP_017922604.1 | 11 - 12 | 1180 | 91 | ||
| XM_018067116.1 | XP_017922605.1 | 10 - 11 | 1102 | 91 | ||
Set Membership: |
|