Search
Search
| Gene Symbol: ASTN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018052684.1 | XP_017908173.1 | 22 - 23 | 3746 | 57 | |
| XM_018052685.1 | XP_017908174.1 | 22 - 23 | 3734 | 57 | ||
| GenBank mRNA | JO644182.2 | - | 23 - 24 | 3540 | 57 | |
| JT661123.1 | - | 2 - 3 | 782 | 57 | ||
Set Membership: |
|