Search
Search
| Gene Symbol: WNK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018048654.1 | XP_017904143.1 | 7 - 8 | 2010 | 56 | |
| XM_018048655.1 | XP_017904144.1 | 7 - 8 | 2010 | 56 | ||
| XM_018048656.1 | XP_017904145.1 | 7 - 8 | 2010 | 56 | ||
| XM_018048657.1 | XP_017904146.1 | 7 - 8 | 2010 | 56 | ||
| XM_018048658.1 | XP_017904147.1 | 7 - 8 | 2010 | 56 | ||
| XM_018048659.1 | XP_017904148.1 | 7 - 8 | 2010 | 56 | ||
| XM_018048660.1 | XP_017904149.1 | 7 - 8 | 2010 | 56 | ||
| XM_018048661.1 | XP_017904150.1 | 7 - 8 | 2010 | 56 | ||
| XM_018048662.1 | XP_017904151.1 | 7 - 8 | 2010 | 56 | ||
| XM_018048663.1 | XP_017904152.1 | 7 - 8 | 2010 | 56 | ||
| XM_018048664.1 | XP_017904153.1 | 7 - 8 | 2010 | 56 | ||
| XM_018048665.1 | XP_017904154.1 | 4 - 5 | 1021 | 56 | ||
| XM_018048666.1 | XP_017904155.1 | 7 - 8 | 2059 | 56 | ||
| XM_018048667.1 | XP_017904156.1 | 7 - 8 | 2059 | 56 | ||
| XM_018048668.1 | XP_017904157.1 | 7 - 8 | 2059 | 56 | ||
| XM_018048669.1 | XP_017904158.1 | 4 - 5 | 1018 | 56 | ||
| GenBank mRNA | JO424233.1 | - | 21 - 22 | 7460 | 56 | |
Set Membership: |
|