Search
Search

SHROOM1| Gene Symbol: SHROOM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018050400.1 | XP_017905889.1 | 4 - 5 | 2563 | 59 | |
| XM_018050401.1 | XP_017905890.1 | 6 - 7 | 2080 | 59 | ||
| XM_018050402.1 | XP_017905891.1 | 6 - 7 | 2028 | 59 | ||
Set Membership: |
|