Search
Search

SLC4A9| Gene Symbol: SLC4A9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018050283.1 | XP_017905772.1 | 6 - 7 | 918 | 56 | |
| XM_018050284.1 | XP_017905773.1 | 6 - 7 | 920 | 56 | ||
| XM_018050285.1 | XP_017905774.1 | 6 - 7 | 923 | 56 | ||
| XM_018050286.1 | XP_017905775.1 | 6 - 7 | 923 | 56 | ||
| XM_018050287.1 | XP_017905776.1 | 6 - 7 | 920 | 56 | ||
| XM_018050288.1 | XP_017905777.1 | 6 - 7 | 919 | 56 | ||
| GenBank mRNA | JT659763.1 | - | 6 - 7 | 834 | 56 | |
Set Membership: |
|