Search
Search

ICOSLG| Gene Symbol: ICOSLG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018051877.1 | XP_017907366.1 | 3 - 4 | 579 | 90 | |
| XM_018051883.1 | XP_017907372.1 | 2 - 3 | 538 | 90 | ||
| XM_018051888.1 | XP_017907377.1 | 3 - 4 | 579 | 90 | ||
Set Membership: |
|