Search
Search

TNRC6B| Gene Symbol: TNRC6B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013964362.2 | XP_013819816.1 | 5 - 6 | 3297 | 60 | |
| XM_018048799.1 | XP_017904288.1 | 8 - 9 | 3157 | 60 | ||
| XM_018048800.1 | XP_017904289.1 | 8 - 9 | 3270 | 60 | ||
| XM_018048801.1 | XP_017904290.1 | 6 - 7 | 3105 | 60 | ||
| XM_018048802.1 | XP_017904291.1 | 7 - 8 | 3247 | 60 | ||
| GenBank mRNA | JT662706.1 | - | 10 - 11 | 3404 | 60 | |
Set Membership: |
|