Search
Search

GABBR2| Gene Symbol: GABBR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005683953.3 | XP_005684010.3 | 3 - 4 | 859 | 58 | |
| XM_018052222.1 | XP_017907711.1 | 3 - 4 | 535 | 58 | ||
Set Membership: |
|