Search
Search

CRABP1| Gene Symbol: CRABP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005695096.3 | XP_005695153.3 | 3 - 4 | 652 | 81 | |
| GenBank mRNA | JO514400.2 | - | 1 - 2 | 288 | 81 | |
| JO619035.2 | - | 1 - 2 | 277 | 81 | ||
| KA305688.1 | - | 3 - 4 | 530 | 81 | ||
Set Membership: |
|