Search
Search
| Gene Symbol: UTRN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013966586.2 | XP_013822040.2 | 43 - 44 | 6707 | 127 | |
| XM_018053299.1 | XP_017908788.1 | 42 - 43 | 6473 | 127 | ||
| XM_018053300.1 | XP_017908789.1 | 43 - 44 | 6410 | 127 | ||
| XM_018053301.1 | XP_017908790.1 | 43 - 44 | 6394 | 127 | ||
| XM_018053302.1 | XP_017908791.1 | 43 - 44 | 6381 | 127 | ||
| XM_018053303.1 | XP_017908792.1 | 42 - 43 | 6474 | 127 | ||
| XM_018053304.1 | XP_017908793.1 | 42 - 43 | 6566 | 127 | ||
| XM_018053305.1 | XP_017908794.1 | 42 - 43 | 6474 | 127 | ||
Set Membership: |
|