Search
Search

CDC14B| Gene Symbol: CDC14B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005684149.3 | XP_005684206.1 | 4 - 5 | 1225 | 83 | |
| XM_018052484.1 | XP_017907973.1 | 4 - 5 | 864 | 83 | ||
| XM_018052485.1 | XP_017907974.1 | 4 - 5 | 865 | 83 | ||
| XM_018052486.1 | XP_017907975.1 | 4 - 5 | 522 | 83 | ||
| XM_018052487.1 | XP_017907976.1 | 4 - 5 | 522 | 83 | ||
| XM_018052488.1 | XP_017907977.1 | 4 - 5 | 522 | 83 | ||
Set Membership: |
|