Search
Search
| Gene Symbol: PBLD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005699113.3 | XP_005699170.1 | 6 - 7 | 616 | 66 | |
| XM_005699114.3 | XP_005699171.1 | 5 - 6 | 942 | 66 | ||
| XM_013975622.2 | XP_013831076.1 | 7 - 8 | 753 | 66 | ||
| XM_013975623.2 | XP_013831077.1 | 7 - 8 | 748 | 66 | ||
| XM_018042229.1 | XP_017897718.1 | 6 - 7 | 541 | 66 | ||
| XM_018042230.1 | XP_017897719.1 | 6 - 7 | 1041 | 66 | ||
Set Membership: |
|