Search
Search
| Gene Symbol: LRP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018059128.1 | XP_017914617.1 | 11 - 12 | 1416 | 54 | |
| XM_018059129.1 | XP_017914618.1 | 11 - 12 | 1418 | 54 | ||
| XM_018059130.1 | XP_017914619.1 | 11 - 12 | 1418 | 54 | ||
| GenBank mRNA | JO424309.1 | - | 27 - 28 | 6990 | 54 | |
| KA317716.1 | - | 4338 | 54 | |||
Set Membership: |
|