Search
Search

CEP104| Gene Symbol: CEP104 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018060459.1 | XP_017915948.1 | 15 - 16 | 2357 | 57 | |
| GenBank mRNA | JO427650.1 | - | 15 - 16 | 2359 | 57 | |
Set Membership: |
|