Search
Search
| Gene Symbol: CEP97 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018061452.1 | XP_017916941.1 | 6 - 7 | 815 | 77 | |
| GenBank mRNA | JO325015.2 | - | 1 - 2 | 63 | 77 | |
| JO395737.2 | - | 1 - 2 | 127 | 77 | ||
| JO443406.2 | - | 6 - 7 | 760 | 77 | ||
| JO544599.2 | - | 1 - 2 | 181 | 77 | ||
| JO596258.2 | - | 5 - 6 | 1330 | 77 | ||
| JO647034.2 | - | 3 - 4 | 939 | 77 | ||
| JT738632.1 | - | 2 - 3 | 244 | 77 | ||
Set Membership: |
|