Search
Search
| Gene Symbol: IFT57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JO355846.2 | - | 7 - 8 | 697 | 61 | |
| JO371531.2 | - | 3 - 4 | 147 | 61 | ||
| JO415608.2 | - | 3 - 4 | 326 | 61 | ||
| JO433624.2 | - | 3 - 4 | 555 | 61 | ||
| JO440478.2 | - | 3 - 4 | 555 | 61 | ||
| JO507482.2 | - | 6 - 7 | 587 | 61 | ||
| JO540780.2 | - | 5 - 6 | 492 | 61 | ||
| JO563030.2 | - | 3 - 4 | 318 | 61 | ||
| JO563246.2 | - | 3 - 4 | 318 | 61 | ||
| JO594341.2 | - | 3 - 4 | 314 | 61 | ||
| JO594342.2 | - | 3 - 4 | 314 | 61 | ||
| JO638726.2 | - | 3 - 4 | 341 | 61 | ||
| KA321683.1 | - | 3 - 4 | 431 | 61 | ||
Set Membership: |
|