Search
Search

B4GALNT1| Gene Symbol: B4GALNT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005680275.3 | XP_005680332.1 | 6 - 7 | 1953 | 62 | |
| XM_005680276.3 | XP_005680333.1 | 6 - 7 | 1953 | 62 | ||
| GenBank mRNA | JO629451.2 | - | 5 - 6 | 1229 | 62 | |
Set Membership: |
|