Search
Search
| Gene Symbol: FEZ2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018055114.1 | XP_017910603.1 | 4 - 5 | 665 | 71 | |
| XM_018055115.1 | XP_017910604.1 | 4 - 5 | 665 | 71 | ||
| XM_018055117.1 | XP_017910606.1 | 3 - 4 | 548 | 71 | ||
| GenBank mRNA | JT679916.1 | - | 7 - 8 | 1147 | 71 | |
Set Membership: |
|