Search
Search

ZC3HAV1| Gene Symbol: ZC3HAV1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018046885.1 | XP_017902374.1 | 9 - 10 | 2569 | 62 | |
| GenBank mRNA | JO595665.2 | - | 4 - 5 | 1953 | 62 | |
Set Membership: |
|