Search
Search

LRRFIP1| Gene Symbol: LRRFIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005678768.3 | XP_005678825.1 | 6 - 7 | 802 | 78 | |
| XM_018040511.1 | XP_017896000.1 | 18 - 19 | 1350 | 78 | ||
| XM_018040600.1 | XP_017896089.1 | 9 - 10 | 843 | 78 | ||
| XM_018040607.1 | XP_017896096.1 | 9 - 10 | 974 | 78 | ||
| XM_018040614.1 | XP_017896103.1 | 8 - 9 | 771 | 78 | ||
| XM_018040623.1 | XP_017896112.1 | 8 - 9 | 901 | 78 | ||
| XM_018040631.1 | XP_017896120.1 | 6 - 7 | 674 | 78 | ||
| XM_018040638.1 | XP_017896127.1 | 6 - 7 | 804 | 78 | ||
| GenBank mRNA | JO578093.1 | - | 6 - 7 | 876 | 78 | |
Set Membership: |
|