Search
Search
| Gene Symbol: BRAF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018046857.1 | XP_017902346.1 | 4 - 5 | 792 | 61 | |
| XM_018046858.1 | XP_017902347.1 | 4 - 5 | 792 | 61 | ||
| XM_018046859.1 | XP_017902348.1 | 4 - 5 | 792 | 61 | ||
| GenBank mRNA | KA352883.1 | - | 4 - 5 | 584 | 61 | |
Set Membership: |
|