Search
Search

HSP90B1| Gene Symbol: HSP90B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018048184.1 | XP_017903673.1 | 11 - 12 | 1549 | 69 | |
| GenBank mRNA | KA321034.1 | - | 11 - 12 | 1573 | 69 | |
Set Membership: |
|