Search
Search
| Gene Symbol: ACBD7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018057140.1 | XP_017912629.1 | 2 - 3 | 211 | 95 | |
| GenBank mRNA | JO632001.2 | - | 2 - 3 | 1899 | 95 | |
| JO632003.2 | - | 2 - 3 | 1897 | 95 | ||
Set Membership: |
|