Search
Search
| Gene Symbol: SEZ6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018064332.1 | XP_017919821.1 | 9 - 10 | 2066 | 55 | |
| XM_018064333.1 | XP_017919822.1 | 9 - 10 | 2066 | 55 | ||
| GenBank mRNA | JO622292.2 | - | 8 - 9 | 2087 | 55 | |
| KA342167.1 | - | 8 - 9 | 2122 | 55 | ||
Set Membership: |
|