Search
Search

IGFLR1| Gene Symbol: IGFLR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005692296.3 | XP_005692353.2 | 2 - 3 | 1027 | 95 | |
| GenBank mRNA | JO640835.2 | - | 1 - 2 | 379 | 95 | |
| JO640836.2 | - | 1 - 2 | 379 | 95 | ||
Set Membership: |
|