Search
Search
| Gene Symbol: CAPSL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005694801.1 | XP_005694858.1 | 3 - 4 | 413 | 101 | |
| XM_018065802.1 | XP_017921291.1 | 3 - 4 | 413 | 101 | ||
| XM_018065803.1 | XP_017921292.1 | 3 - 4 | 413 | 101 | ||
Set Membership: |
|