Search
Search

PDCD1LG2| Gene Symbol: PDCD1LG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013965947.2 | XP_013821401.2 | 3 - 4 | 1105 | 80 | |
| XM_013965949.2 | XP_013821403.2 | 3 - 4 | 1101 | 80 | ||
Set Membership: |
|