Search
Search

DEPDC5| Gene Symbol: DEPDC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018060922.1 | XP_017916411.1 | 21 - 22 | 1918 | 58 | |
| XM_018060923.1 | XP_017916412.1 | 21 - 22 | 1918 | 58 | ||
| XM_018060924.1 | XP_017916413.1 | 21 - 22 | 1918 | 58 | ||
| XM_018060925.1 | XP_017916414.1 | 21 - 22 | 1918 | 58 | ||
| GenBank mRNA | JO594185.2 | - | 10 - 11 | 1384 | 58 | |
Set Membership: |
|