Search
Search
| Gene Symbol: TRAF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018040663.1 | XP_017896152.1 | 20 - 21 | 2147 | 113 | |
| XM_018040664.1 | XP_017896153.1 | 20 - 21 | 2350 | 113 | ||
| XM_018040665.1 | XP_017896154.1 | 20 - 21 | 2141 | 113 | ||
| GenBank mRNA | JO332375.2 | - | 13 - 14 | 1447 | 113 | |
Set Membership: |
|