Search
Search

C17H4orf33| Gene Symbol: C17H4orf33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005691256.3 | XP_005691313.1 | 3 - 4 | 327 | 113 | |
| XM_018061531.1 | XP_017917020.1 | 3 - 4 | 377 | 113 | ||
| XM_018061532.1 | XP_017917021.1 | 3 - 4 | 292 | 113 | ||
| GenBank mRNA | JO453588.2 | - | 3 - 4 | 300 | 113 | |
Set Membership: |
|