Search
Search

ZNF75D| Gene Symbol: ZNF75D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005700407.3 | XP_005700464.1 | 5 - 6 | 1207 | 89 | |
| XM_005700408.3 | XP_005700465.1 | 5 - 6 | 1047 | 89 | ||
| XM_018044138.1 | XP_017899627.1 | 5 - 6 | 1180 | 89 | ||
| XM_018044139.1 | XP_017899628.1 | 5 - 6 | 1215 | 89 | ||
| XM_018044140.1 | XP_017899629.1 | 5 - 6 | 1214 | 89 | ||
| XM_018044141.1 | XP_017899630.1 | 5 - 6 | 1213 | 89 | ||
| XM_018044142.1 | XP_017899631.1 | 5 - 6 | 1212 | 89 | ||
| GenBank mRNA | JO496211.2 | - | 1060 | 89 | ||
| JO580121.1 | - | 3 - 4 | 692 | 89 | ||
Set Membership: |
|