Search
Search

EPB41L1| Gene Symbol: EPB41L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018057797.1 | XP_017913286.1 | 25 - 26 | 5064 | 60 | |
| XM_018057798.1 | XP_017913287.1 | 24 - 25 | 5012 | 60 | ||
| XM_018057799.1 | XP_017913288.1 | 24 - 25 | 5021 | 60 | ||
| XM_018057800.1 | XP_017913289.1 | 24 - 25 | 5246 | 60 | ||
| XM_018057801.1 | XP_017913290.1 | 24 - 25 | 5328 | 60 | ||
| XM_018057802.1 | XP_017913291.1 | 24 - 25 | 5154 | 60 | ||
| XM_018057803.1 | XP_017913292.1 | 20 - 21 | 2635 | 60 | ||
| XM_018057804.1 | XP_017913293.1 | 20 - 21 | 2632 | 60 | ||
| XM_018057805.1 | XP_017913294.1 | 19 - 20 | 2223 | 60 | ||
| GenBank mRNA | JO424536.1 | - | 1 - 2 | 3404 | 60 | |
Set Membership: |
|