Search
Search

SLITRK2| Gene Symbol: SLITRK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018044268.1 | XP_017899757.1 | 4 | 2903 | 77 | |
| XM_018044269.1 | XP_017899758.1 | 2 | 2715 | 77 | ||
| GenBank mRNA | JO639016.2 | - | 1 | 380 | 77 | |
| JO639017.2 | - | 1 | 380 | 77 | ||
Set Membership: |
|