Search
Search

ATP6V1C2| Gene Symbol: ATP6V1C2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005687003.3 | XP_005687060.1 | 5 - 6 | 774 | 59 | |
| XM_005687004.3 | XP_005687061.1 | 5 - 6 | 771 | 59 | ||
| XM_018055655.1 | XP_017911144.1 | 5 - 6 | 487 | 59 | ||
| XM_018055656.1 | XP_017911145.1 | 5 - 6 | 465 | 59 | ||
| XM_018055657.1 | XP_017911146.1 | 5 - 6 | 488 | 59 | ||
| XM_018055658.1 | XP_017911147.1 | 5 - 6 | 425 | 59 | ||
Set Membership: |
|