Search
Search

TRAPPC13| Gene Symbol: TRAPPC13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005694630.3 | XP_005694687.1 | 9 - 10 | 1280 | 92 | |
| XM_005694631.3 | XP_005694688.1 | 9 - 10 | 1280 | 92 | ||
| XM_005694632.3 | XP_005694689.1 | 8 - 9 | 1262 | 92 | ||
| XM_005694633.3 | XP_005694690.1 | 8 - 9 | 1263 | 92 | ||
| GenBank mRNA | JO395535.2 | - | 7 - 8 | 508 | 92 | |
Set Membership: |
|