Search
Search
| Gene Symbol: EML1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018066339.1 | XP_017921828.1 | 10 - 11 | 1157 | 59 | |
| XM_018066340.1 | XP_017921829.1 | 10 - 11 | 1154 | 59 | ||
| XM_018066341.1 | XP_017921830.1 | 10 - 11 | 1151 | 59 | ||
| XM_018066342.1 | XP_017921831.1 | 10 - 11 | 1198 | 59 | ||
| XM_018066343.1 | XP_017921832.1 | 9 - 10 | 1100 | 59 | ||
| XM_018066344.1 | XP_017921833.1 | 9 - 10 | 1097 | 59 | ||
| XM_018066345.1 | XP_017921834.1 | 9 - 10 | 1137 | 59 | ||
Set Membership: |
|