Search
Search

ZNF576| Gene Symbol: ZNF576 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005692610.3 | XP_005692667.2 | 2 - 3 | 490 | 91 | |
| XM_005692611.3 | XP_005692668.1 | 2 - 3 | 245 | 91 | ||
| GenBank mRNA | JO636767.2 | - | 1 - 2 | 919 | 91 | |
| JO636768.2 | - | 2 - 3 | 550 | 91 | ||
Set Membership: |
|