Search
Search

AGPAT3| Gene Symbol: AGPAT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005675655.3 | XP_005675712.2 | 8 - 9 | 969 | 85 | |
| XM_018051810.1 | XP_017907299.1 | 8 - 9 | 1085 | 85 | ||
| XM_018051819.1 | XP_017907308.1 | 9 - 10 | 1163 | 85 | ||
| GenBank mRNA | JO642969.2 | - | 8 - 9 | 1104 | 85 | |
Set Membership: |
|