Search
Search

CFAP70| Gene Symbol: CFAP70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005699201.3 | XP_005699258.1 | 22 - 23 | 2787 | 64 | |
| GenBank mRNA | JT668177.1 | - | 9 - 10 | 679 | 64 | |
Set Membership: |
|