Search
Search

LOC102171703| Gene Symbol: LOC102171703 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013965428.2 | XP_013820882.2 | 4 - 5 | 1165 | 59 | |
| XM_018050551.1 | XP_017906040.1 | 4 - 5 | 1166 | 59 | ||
| XM_018050552.1 | XP_017906041.1 | 4 - 5 | 1166 | 59 | ||
| GenBank mRNA | JO636161.2 | - | 4 - 5 | 886 | 59 | |
Set Membership: |
|