Search
Search
| Gene Symbol: PKP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018060842.1 | XP_017916331.1 | 4 - 5 | 1087 | 54 | |
| GenBank mRNA | JO366442.2 | - | 3698 | 54 | ||
| KA335660.1 | - | 3706 | 54 | |||
Set Membership: |
|