Search
Search

ADGRB2| Gene Symbol: ADGRB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018057533.1 | XP_017913022.1 | 2 - 3 | 1026 | 61 | |
| XM_018057542.1 | XP_017913031.1 | 2 - 3 | 1026 | 61 | ||
| XM_018057552.1 | XP_017913041.1 | 2 - 3 | 1026 | 61 | ||
| XM_018057563.1 | XP_017913052.1 | 2 - 3 | 1026 | 61 | ||
| XM_018057569.1 | XP_017913058.1 | 2 - 3 | 1026 | 61 | ||
| XM_018057576.1 | XP_017913065.1 | 1 - 2 | 830 | 61 | ||
Set Membership: |
|