Search
Search
| Gene Symbol: FCGRT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018062789.1 | XP_017918278.1 | 5 - 6 | 1000 | 84 | |
| XM_018062790.1 | XP_017918279.1 | 4 - 5 | 1072 | 84 | ||
| GenBank mRNA | JO579294.1 | - | 5 - 6 | 1057 | 84 | |
Set Membership: |
|