Search
Search

HEATR5A| Gene Symbol: HEATR5A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018065895.1 | XP_017921384.1 | 12 - 13 | 2080 | 59 | |
| XM_018065896.1 | XP_017921385.1 | 11 - 12 | 2013 | 59 | ||
| XM_018065897.1 | XP_017921386.1 | 12 - 13 | 2080 | 59 | ||
| XM_018065898.1 | XP_017921387.1 | 12 - 13 | 2077 | 59 | ||
Set Membership: |
|